Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289160.1 | NP_001276089.1 | 2 | 145 | ||
NM_001289161.1 | NP_001276090.1 | 2 | 244 | |||
NM_005931.4 | NP_005922.2 | 2 | 244 | |||
GenBank mRNA | AF021221.1 | - | 1 | 30 | ||
AF021222.1 | - | 1 | 30 | |||
AF021223.1 | - | 1 | 30 | |||
AF021224.1 | - | 1 | 30 | |||
AF021225.1 | - | 1 | 30 | |||
AF021226.1 | - | 1 | 30 | |||
AK291927.1 | - | 2 | 242 | |||
AK300791.1 | - | 2 | 145 | |||
AK303655.1 | - | 2 | 108 | |||
AK314228.1 | - | 2 | 149 | |||
AY885251.1 | - | 3 | 196 | |||
BC044218.1 | - | 2 | 208 | 6025253 | ||
DC364717.1 | - | 2 | 108 | |||
DC427936.1 | - | 2 | 216 | |||
GU235997.1 | - | 2 | 98 | |||
U95729.1 | - | 1 | 31 | |||
U95730.1 | - | 1 | 31 | |||
U95731.1 | - | 1 | 31 | |||
U95732.1 | - | 1 | 31 | |||
U95733.1 | - | 1 | 31 | |||
U95734.1 | - | 1 | 31 | |||
X91625.1 | - | 2 | 105 |