Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286435.1 | NP_001273364.1 | 3 | 337 | ||
NM_001286436.1 | NP_001273365.1 | 3 | 540 | |||
NM_001286438.1 | NP_001273367.1 | 4 | 515 | |||
NM_001286439.1 | NP_001273368.1 | 4 | 524 | |||
NM_001286440.1 | NP_001273369.1 | 3 | 653 | |||
NM_005009.2 | NP_005000.1 | 3 | 337 | |||
GenBank mRNA | AF086133.1 | - | 1 | 35 | ||
AK094439.1 | - | 1 | 1050 | |||
BC004880.2 | - | 3 | 320 | 3830205 | ||
BC017067.1 | - | 3 | 337 | 3913604 | ||
BE266496.1 | - | 3 | 292 | |||
BP328820.1 | - | 4 | 410 | |||
BQ650649.1 | - | 3 | 496 | |||
BQ932845.1 | - | 3 | 591 | |||
BT019438.1 | - | 3 | 306 | |||
CF126980.1 | - | 4 | 489 | |||
Y07604.1 | - | 3 | 317 |