Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NUMA1 SIRNA Assays ›
Gene Symbol:
NUMA1
Gene Name:
nuclear mitotic apparatus protein 1
Gene Aliases:
NMP-22, NUMA
Chromosome Location:
Chr.11: 72002864 - 72080693 on Build GRCh38
UniGene ID:
Hs.325978
Species:
Human
siRNA ID
HSS181605
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NUMA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286561.1
NP_001273490.1
7, 8
733
NM_006185.3
NP_006176.2
6, 7
635
NR_104476.1
6, 7
554
XM_006718564.1
XP_006718627.1
5, 6
450
XM_011545054.1
XP_011543356.1
5, 6
450
XM_011545055.1
XP_011543357.1
6, 7
628
XM_011545056.2
XP_011543358.1
7, 8
622
XM_011545057.2
XP_011543359.1
7, 8
612
XM_011545058.1
XP_011543360.1
5, 6
649
XM_011545059.2
XP_011543361.1
7, 8
614
XM_011545060.2
XP_011543362.1
7, 8
608
XM_011545061.2
XP_011543363.1
5, 6
468
XM_011545062.2
XP_011543364.1
5, 6
481
XM_011545063.2
XP_011543365.1
5, 6
448
XM_011545064.1
XP_011543366.1
5, 6
435
XM_011545065.1
XP_011543367.1
5, 6
450
XM_011545066.1
XP_011543368.1
5, 6
450
XM_017017830.1
XP_016873319.1
6, 7
490
XM_017017831.1
XP_016873320.1
5, 6
486
GenBank mRNA
AB210007.1
-
6, 7
436
AB621824.1
-
7, 8
617
AK290509.1
-
5, 6
450
AK307577.1
-
6, 7
453
AK309041.1
-
5, 6
457
BC004165.2
-
6, 7
469
BC027493.1
-
6, 7
436
BC043499.1
-
6, 7
436
BC103765.1
-
6, 7
554
Z11583.1
-
6, 7
453
Z11584.1
-
7, 8
555
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
establishment of mitotic spindle orientation
nucleus organization
mitotic nuclear division
cell division
meiotic cell cycle
lung epithelial cell differentiation
structural molecule activity
protein binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0