Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024822.2 | NP_001019993.1 | 1 | 275 | ||
GenBank mRNA | AW274139.1 | - | 1 | 470 | ||
AY665807.1 | - | 1 | 175 | |||
BC130347.1 | - | 1 | 274 | 40146378 | ||
BC130349.1 | - | 1 | 275 | 40146380 | ||
FJ237362.1 | - | 1 | 275 |
Gene Symbol: LOC254028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122043.1 | 2 | 204 | |||
GenBank mRNA | AK093210.1 | - | 1 | 144 | ||
BI830942.1 | - | 2 | 204 | |||
DB053670.1 | - | 2 | 275 |