Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ODF2 SIRNA Assays ›
Gene Symbol:
ODF2
Gene Name:
outer dense fiber of sperm tails 2
Gene Aliases:
CT134, ODF2/1, ODF2/2, ODF84
Chromosome Location:
Chr.9: 128455155 - 128501292 on Build GRCh38
UniGene ID:
Hs.129055
Species:
Human
siRNA ID
HSS181634
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ODF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242352.1
NP_001229281.1
13, 14
1777
NM_001242353.1
NP_001229282.1
13, 14
1863
NM_001242354.1
NP_001229283.1
12, 13
1549
NM_002540.4
NP_002531.3
15, 16
2065
NM_153432.1
NP_702910.1
12, 13
1623
NM_153433.1
NP_702911.1
13, 14
1790
NM_153435.1
NP_702913.1
13, 14
1683
NM_153436.1
NP_702914.1
12, 13
1447
NM_153437.2
NP_702915.1
13, 14
1713
NM_153439.1
NP_702917.1
12, 13
1566
NM_153440.1
NP_702918.1
12, 13
1493
XM_005252010.2
XP_005252067.1
12, 13
1575
XM_005252011.2
XP_005252068.1
12, 13
1515
XM_005252012.3
XP_005252069.1
13, 14
1765
XM_005252013.3
XP_005252070.1
13, 14
1712
XM_005252014.3
XP_005252071.1
13, 14
1761
XM_005252017.4
XP_005252074.1
13, 14
1666
XM_005252018.2
XP_005252075.1
14, 15
1871
XM_005252019.3
XP_005252076.1
14, 15
1890
XM_005252022.2
XP_005252079.1
13, 14
1689
XM_005252023.2
XP_005252080.1
13, 14
1660
XM_005252024.3
XP_005252081.1
13, 14
1679
XM_005252026.4
XP_005252083.1
13, 14
1608
XM_005252029.2
XP_005252086.1
13, 14
1689
XM_006717126.3
XP_006717189.1
13, 14
1765
XM_011518722.2
XP_011517024.1
13, 14
1689
XM_011518723.2
XP_011517025.1
13, 14
1632
XM_011518724.2
XP_011517026.1
14, 15
1853
XM_011518725.2
XP_011517027.1
14, 15
1857
XM_011518726.2
XP_011517028.1
14, 15
1875
XM_011518728.2
XP_011517030.1
13, 14
1626
XM_011518729.2
XP_011517031.1
13, 14
1562
XM_011518730.2
XP_011517032.1
13, 14
1689
XM_011518731.2
XP_011517033.1
12, 13
1518
XM_011518732.2
XP_011517034.1
13, 14
1475
XM_011518733.2
XP_011517035.1
12, 13
1461
XM_011518744.1
XP_011517046.1
12, 13
1515
XM_017014764.1
XP_016870253.1
14, 15
1846
XM_017014765.1
XP_016870254.1
14, 15
1865
XM_017014766.1
XP_016870255.1
12, 13
1575
XM_017014767.1
XP_016870256.1
14, 15
1857
XM_017014768.1
XP_016870257.1
13, 14
1649
XM_017014769.1
XP_016870258.1
13, 14
1678
XM_017014770.1
XP_016870259.1
14, 15
1833
XM_017014771.1
XP_016870260.1
14, 15
1814
XM_017014772.1
XP_016870261.1
14, 15
1890
XM_017014773.1
XP_016870262.1
14, 15
1866
XM_017014774.1
XP_016870263.1
13, 14
1576
XM_017014775.1
XP_016870264.1
14, 15
1833
XM_017014776.1
XP_016870265.1
14, 15
1814
XM_017014777.1
XP_016870266.1
13, 14
1616
XM_017014778.1
XP_016870267.1
13, 14
1719
XM_017014779.1
XP_016870268.1
12, 13
1460
XM_017014780.1
XP_016870269.1
13, 14
1643
XM_017014781.1
XP_016870270.1
12, 13
1398
GenBank mRNA
AF012549.1
-
13, 14
1653
AF053970.1
-
11, 12
1352
AK093367.1
-
12, 13
1573
AK299303.1
-
13, 14
1625
AK300793.1
-
12, 13
1398
AK301754.1
-
12, 13
1490
AK301842.1
-
12, 13
1415
AK301855.1
-
12, 13
1469
AK302684.1
-
13, 14
1717
AY319414.1
-
12, 13
1599
AY366499.1
-
13, 14
1560
BC010629.1
-
15, 16
1863
3874501
BC091500.1
-
13, 14
1677
30378678
DQ444713.1
-
11, 12
1390
DQ444714.1
-
11, 12
1333
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
multicellular organism development
spermatid development
structural molecule activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0