Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006198.2 | NP_006189.2 | 3 | 193 | ||
GenBank mRNA | AF416716.1 | - | 3 | 141 | ||
AK289964.1 | - | 3 | 176 | |||
AM183336.1 | - | 3 | 131 | |||
BC013791.1 | - | 3 | 169 | 4345740 | ||
BQ287895.1 | - | 1 | 370 | |||
CR450319.1 | - | 2 | 112 | |||
U52969.1 | - | 3 | 193 | |||
X93349.1 | - | 3 | 170 |