Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GULP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252668.1 | NP_001239597.1 | 7 | 1018 | ||
NM_016315.3 | NP_057399.1 | 7 | 1018 | |||
NR_045562.1 | 7 | 970 | ||||
NR_045563.1 | 8 | 1232 | ||||
XM_006712580.3 | XP_006712643.1 | 7 | 1084 | |||
XM_006712581.1 | XP_006712644.1 | 7 | 978 | |||
XM_006712582.1 | XP_006712645.1 | 7 | 1021 | |||
XM_006712583.3 | XP_006712646.1 | 8 | 1300 | |||
XM_006712584.3 | XP_006712647.1 | 7 | 1174 | |||
XM_006712585.1 | XP_006712648.1 | 6 | 487 | |||
XM_006712589.3 | XP_006712652.1 | 7 | 978 | |||
XM_011511329.1 | XP_011509631.1 | 7 | 978 | |||
XM_011511331.1 | XP_011509633.1 | 5 | 495 | |||
XM_011511332.1 | XP_011509634.1 | 8 | 1089 | |||
XM_017004303.1 | XP_016859792.1 | 7 | 978 | |||
XM_017004304.1 | XP_016859793.1 | 7 | 1084 | |||
XM_017004305.1 | XP_016859794.1 | 7 | 1021 | |||
XM_017004306.1 | XP_016859795.1 | 7 | 1350 | |||
GenBank mRNA | AF191771.1 | - | 7 | 777 | ||
AF200715.1 | - | 7 | 783 | |||
AK023668.1 | - | 7 | 1096 | |||
AK055718.1 | - | 7 | 1018 | |||
AK314498.1 | - | 7 | 812 | |||
BC001103.2 | - | 7 | 741 | 3505407 |