Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFKP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242339.1 | NP_001229268.1 | 17 | 1700 | ||
NM_001323067.1 | NP_001309996.1 | 15 | 1439 | |||
NM_001323068.1 | NP_001309997.1 | 15 | 1576 | |||
NM_001323069.1 | NP_001309998.1 | 15 | 1571 | |||
NM_001323070.1 | NP_001309999.1 | 10 | 1386 | |||
NM_001323071.1 | NP_001310000.1 | 16 | 1692 | |||
NM_001323072.1 | NP_001310001.1 | 16 | 1799 | |||
NM_001323073.1 | NP_001310002.1 | 11 | 1060 | |||
NM_001323074.1 | NP_001310003.1 | 10 | 1386 | |||
NM_002627.4 | NP_002618.1 | 15 | 1576 | |||
XM_005252465.4 | XP_005252522.1 | 15 | 1621 | |||
XM_005252466.4 | XP_005252523.1 | 15 | 1621 | |||
XM_005252467.2 | XP_005252524.1 | 15 | 1545 | |||
XM_006717449.1 | XP_006717512.1 | 15 | 1545 | |||
GenBank mRNA | AK092597.1 | - | 17 | 1700 | ||
AK128373.1 | - | 2 | 2088 | |||
AK291841.1 | - | 15 | 1548 | |||
AK308003.1 | - | 15 | 1539 | |||
AK308226.1 | - | 11 | 976 | |||
AK309692.1 | - | 8 | 876 | |||
BC002536.1 | - | 15 | 1536 | 3140233 | ||
BC021698.1 | - | 6 | 489 | |||
BC029138.1 | - | 15 | 1576 | 5180268 | ||
D21863.1 | - | 5 | 386 | |||
D25328.1 | - | 15 | 1533 |