Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201564.1 | NP_001188493.1 | 9 | 879 | ||
NM_002669.3 | NP_002660.1 | 9 | 906 | |||
GenBank mRNA | AF044333.1 | - | 8 | 781 | ||
AK002108.1 | - | 9 | 881 | |||
AK304574.1 | - | 8 | 774 | |||
AK307997.1 | - | 8 | 655 | |||
BC020786.1 | - | 9 | 843 | 4809482 | ||
BC106004.1 | - | 9 | 844 | 3889109 |