Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019005.3 | NP_061878.3 | 11 | 2669 | ||
XM_005249780.3 | XP_005249837.1 | 10 | 2697 | |||
XM_005249781.3 | XP_005249838.1 | 9 | 2513 | |||
XM_005249782.3 | XP_005249839.1 | 9 | 2898 | |||
XM_011515432.2 | XP_011513734.1 | 7 | 2183 | |||
XM_017012361.1 | XP_016867850.1 | 10 | 2864 | |||
XM_017012362.1 | XP_016867851.1 | 10 | 2703 | |||
GenBank mRNA | AK000330.1 | - | 10 | 2538 | ||
AK294561.1 | - | 11 | 2628 | |||
AL136892.1 | - | 6 | 1132 | |||
BC005883.2 | - | 8 | 1348 | |||
BC140833.1 | - | 10 | 2519 | 9021701 | ||
BC140834.1 | - | 9 | 2421 | 9021702 |