Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POU6F1 SIRNA Assays ›
Gene Symbol:
POU6F1
Gene Name:
POU class 6 homeobox 1
Gene Aliases:
BRN5, MPOU, TCFB1
Chromosome Location:
Chr.12: 51186936 - 51218134 on Build GRCh38
UniGene ID:
Hs.555886
Species:
Human
siRNA ID
HSS182595
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POU6F1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002702.3
NP_002693.3
6
1073
NR_026893.1
11
1531
XM_017019507.1
XP_016874996.1
11
2718
XM_017019508.1
XP_016874997.1
12
2629
XM_017019509.1
XP_016874998.1
10
1638
XM_017019510.1
XP_016874999.1
11
2067
XM_017019511.1
XP_016875000.1
11
2236
XM_017019512.1
XP_016875001.1
11
1592
XM_017019513.1
XP_016875002.1
11
2715
XM_017019514.1
XP_016875003.1
11
2712
XM_017019515.1
XP_016875004.1
11
2058
XM_017019516.1
XP_016875005.1
11
2709
XM_017019517.1
XP_016875006.1
10
2589
XM_017019518.1
XP_016875007.1
10
2583
XM_017019519.1
XP_016875008.1
10
2543
XM_017019520.1
XP_016875009.1
9
1945
XM_017019521.1
XP_016875010.1
9
1964
XM_017019522.1
XP_016875011.1
10
1442
XM_017019523.1
XP_016875012.1
8
1149
XM_017019524.1
XP_016875013.1
10
1540
XM_017019525.1
XP_016875014.1
11
1528
XM_017019526.1
XP_016875015.1
10
1436
XM_017019527.1
XP_016875016.1
10
1455
XM_017019528.1
XP_016875017.1
11
1961
XM_017019529.1
XP_016875018.1
10
1847
XM_017019530.1
XP_016875019.1
9
1752
GenBank mRNA
AK095191.1
-
11
1531
AL832881.1
-
6
1073
BC051326.1
-
6
876
6275848
BC074765.2
-
5
616
30915178
L14482.1
-
6
744
Z21966.1
-
5
783
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
heart development
muscle organ development
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0