Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYTL2 SIRNA Assays ›
Gene Symbol:
SYTL2
Gene Name:
synaptotagmin like 2
Gene Aliases:
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location:
Chr.11: 85694221 - 85811182 on Build GRCh38
UniGene ID:
Hs.369520
Species:
Human
siRNA ID
HSS182677
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYTL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162951.2
NP_001156423.1
14
2515
NM_001162952.2
NP_001156424.1
6
1045
NM_001162953.2
NP_001156425.1
14
2518
NM_001289608.1
NP_001276537.1
15
2324
NM_001289609.1
NP_001276538.1
7
1158
NM_001289610.1
NP_001276539.1
7
729
NM_032943.4
NP_116561.1
15
2694
NM_206929.3
NP_996812.1
8
849
NM_206930.3
NP_996813.1
5
1022
XM_005274057.1
XP_005274114.1
14
6154
XM_005274060.1
XP_005274117.1
15
2338
XM_005274061.1
XP_005274118.1
15
2335
XM_005274062.2
XP_005274119.1
16
2759
XM_005274063.1
XP_005274120.1
14
2218
XM_005274066.1
XP_005274123.1
13
2167
XM_005274067.4
XP_005274124.1
7
2461
XM_011545105.1
XP_011543407.1
15
6202
XM_011545106.1
XP_011543408.1
15
6199
XM_011545107.2
XP_011543409.1
16
6623
XM_011545108.1
XP_011543410.1
14
6082
XM_011545109.2
XP_011543411.1
15
6184
XM_011545114.2
XP_011543416.1
6
2367
XM_011545115.1
XP_011543417.1
4
461
XM_011545116.2
XP_011543418.1
5
2246
XM_017017933.1
XP_016873422.1
14
6151
XM_017017934.1
XP_016873423.1
16
6620
XM_017017935.1
XP_016873424.1
12
5998
XM_017017936.1
XP_016873425.1
12
5696
XM_017017937.1
XP_016873426.1
15
2239
XM_017017938.1
XP_016873427.1
14
2197
XM_017017939.1
XP_016873428.1
8
2505
XM_017017940.1
XP_016873429.1
7
2457
GenBank mRNA
AB046817.1
-
15
2705
AK024872.1
-
5
1186
AK074737.1
-
8
806
AK092121.1
-
5
840
AK124754.1
-
8
2430
AK127504.1
-
9
3393
AK131365.1
-
8
1322
AK222707.1
-
5
1186
AK222710.1
-
5
1186
AK296224.1
-
15
2306
AK296740.1
-
12
2254
AK298604.1
-
7
647
AL601023.1
-
7
702
AL834422.1
-
2
84
AY386362.1
-
8
4709
BC015540.2
-
5
516
BC068495.1
-
7
883
BC110315.1
-
6
574
BC136450.1
-
14
2450
9020439
BC144114.1
-
14
2450
9052634
BC144115.1
-
14
2447
9052635
BX647671.1
-
9
3145
BX648068.1
-
8
2591
EU035829.1
-
14
2171
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
vesicle docking involved in exocytosis
vesicle fusion
negative regulation of phosphatase activity
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of mucus secretion
protein localization to plasma membrane
phosphatidylserine binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
Rab GTPase binding
phosphatase binding
syntaxin binding
clathrin binding
neurexin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0