Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184782.1 | NP_001171711.1 | 1 | 681 | ||
NM_018015.5 | NP_060485.4 | 1 | 681 | |||
GenBank mRNA | AK001040.1 | - | 1 | 636 | ||
AK291730.1 | - | 1 | 630 | |||
BC011483.1 | - | 1 | 602 | 3453700 | ||
BC033709.1 | - | 1 | 593 | 3910777 | ||
BC054032.2 | - | 1 | 569 | |||
BC070110.1 | - | 1 | 632 | 5264812 |