Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181575.4 | NP_853553.1 | 11 | 1389 | ||
NR_126510.1 | 11 | 1451 | ||||
NR_126511.1 | 10 | 1585 | ||||
XM_005264392.3 | XP_005264449.1 | 10 | 1334 | |||
GenBank mRNA | AF100746.1 | - | 11 | 1162 | ||
AF100753.1 | - | 11 | 1189 | |||
AF100754.1 | - | 10 | 1387 | |||
AF165515.1 | - | 11 | 1169 | |||
AI580873.1 | - | 2 | 268 | |||
AK023983.1 | - | 11 | 1229 | |||
AK055566.1 | - | 7 | 2021 | |||
AK293121.1 | - | 11 | 1208 | |||
AK304886.1 | - | 10 | 1331 | |||
BC001658.1 | - | 11 | 1314 | 3050204 | ||
BC033646.2 | - | 11 | 1204 | |||
CR749661.1 | - | 10 | 1322 |