Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142627.2 | NP_001136099.1 | 9 | 1044 | ||
NM_001142628.1 | NP_001136100.1 | 8 | 978 | |||
NM_018144.3 | NP_060614.2 | 9 | 1044 | |||
NR_024576.2 | 9 | 1044 | ||||
NR_024577.2 | 10 | 1177 | ||||
GenBank mRNA | AF346603.1 | - | 9 | 985 | ||
AK001440.1 | - | 2 | 279 | |||
AK022027.1 | - | 3 | 427 | |||
AK022640.1 | - | 9 | 979 | |||
AK057532.1 | - | 10 | 1118 | |||
AK292295.1 | - | 9 | 994 | |||
AK301841.1 | - | 8 | 931 | |||
BC026179.1 | - | 9 | 994 | 4823130 |