Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012329.1 | NP_001012329.1 | 3, 4 | 361 | ||
NM_020248.2 | NP_064633.1 | 4, 5 | 395 | |||
XM_017001853.1 | XP_016857342.1 | 4, 5 | 620 | |||
XM_017001854.1 | XP_016857343.1 | 4, 5 | 494 | |||
XM_017001855.1 | XP_016857344.1 | 3, 4 | 337 | |||
GenBank mRNA | AB021262.1 | - | 4, 5 | 361 | ||
BC014300.1 | - | 3, 4 | 349 | |||
BC146892.1 | - | 1, 2 | 124 | 9056820 | ||
BC146896.1 | - | 1, 2 | 124 | 9056824 | ||
BI226022.1 | - | 348 | ||||
CB995652.1 | - | 388 |