Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270427.1 | NP_001257356.1 | 5 | 729 | ||
NM_005047.3 | NP_005038.1 | 6 | 858 | |||
XM_011518866.2 | XP_011517168.1 | 6 | 768 | |||
XM_017014927.1 | XP_016870416.1 | 6 | 768 | |||
XM_017014928.1 | XP_016870417.1 | 5 | 658 | |||
GenBank mRNA | AK026925.1 | - | 2 | 161 | ||
AK291051.1 | - | 6 | 759 | |||
AK303007.1 | - | 5 | 659 | |||
BC014478.1 | - | 6 | 747 | 4878087 | ||
D31889.1 | - | 6 | 745 | |||
S79862.1 | - | 6 | 749 |