Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291941.1 | NP_001278870.1 | 4 | 334 | ||
NM_001291942.1 | NP_001278871.1 | 3 | 222 | |||
NM_001291943.1 | NP_001278872.1 | 4 | 334 | |||
NM_020685.4 | NP_065736.1 | 4 | 441 | |||
XM_017006931.1 | XP_016862420.1 | 4 | 817 | |||
GenBank mRNA | AF236158.1 | - | 4 | 271 | ||
AK310431.1 | - | 4 | 412 | |||
AK313915.1 | - | 4 | 441 | |||
BC017772.2 | - | 3 | 251 | 4307022 | ||
BU568674.1 | - | 176 | ||||
DA615061.1 | - | 4 | 292 | |||
DA783189.1 | - | 4 | 334 |