Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009959.2 | NP_001009959.1 | 4 | 744 | ||
NM_001304344.1 | NP_001291273.1 | 4 | 592 | |||
NM_001304345.1 | NP_001291274.1 | 4 | 604 | |||
NM_001304346.1 | NP_001291275.1 | 3 | 717 | |||
NM_020711.2 | NP_065762.1 | 3 | 777 | |||
GenBank mRNA | AB033015.1 | - | 3 | 519 | ||
AK295051.1 | - | 2 | 452 | |||
AK295844.1 | - | 3 | 593 | |||
AK296477.1 | - | 4 | 687 | |||
AK296489.1 | - | 3 | 338 | |||
BC026345.1 | - | 3 | 603 | 4798869 | ||
DQ334271.1 | - | 3 | 600 |