Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024808.2 | NP_001019979.1 | 6 | 804 | ||
NM_020993.4 | NP_066273.1 | 6 | 867 | |||
GenBank mRNA | AK296159.1 | - | 6 | 806 | ||
AK316486.1 | - | 6 | 702 | |||
BC094723.1 | - | 6 | 615 | 5300018 | ||
BC113627.1 | - | 6 | 691 | 8322679 | ||
X89984.1 | - | 6 | 1614 |