Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080523.1 | NP_001073992.1 | 2 | 444 | ||
XM_005259616.2 | XP_005259673.1 | 2 | 178 | |||
XM_011528181.2 | XP_011526483.1 | 2 | 223 | |||
XM_017027144.1 | XP_016882633.1 | 2 | 658 | |||
XM_017027145.1 | XP_016882634.1 | 2 | 658 |