Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321970.1 | NP_001308899.1 | 2 | 403 | ||
NM_001321972.1 | NP_001308901.1 | 2 | 406 | |||
NM_001321973.1 | NP_001308902.1 | 2 | 403 | |||
NM_001321974.1 | NP_001308903.1 | 2 | 406 | |||
NM_001321976.1 | NP_001308905.1 | 2 | 403 | |||
NM_022839.4 | NP_073750.2 | 2 | 406 | |||
NM_176805.3 | NP_789775.1 | 2 | 406 | |||
NR_135915.1 | 2 | 406 | ||||
NR_135917.1 | 2 | 406 | ||||
NR_135918.1 | 2 | 406 | ||||
GenBank mRNA | AB049944.1 | - | 2 | 406 | ||
AK027059.1 | - | 2 | 152 | |||
AK223152.1 | - | 2 | 151 | |||
AK315407.1 | - | 2 | 151 | |||
BC009451.1 | - | 2 | 138 | |||
BC012489.1 | - | 2 | 160 | 4452949 | ||
BC032378.1 | - | 2 | 175 | 5110633 | ||
BM966864.1 | - | 2 | 141 | |||
CD172722.1 | - | 2 | 172 | |||
DA803430.1 | - | 2 | 154 |