Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000453.2 | NP_000444.1 | 2 | 745 | ||
XM_011528192.2 | XP_011526494.1 | 2 | 1324 | |||
XM_011528193.2 | XP_011526495.1 | 3 | 827 | |||
XM_011528194.2 | XP_011526496.1 | 2 | 232 | |||
XM_017027158.1 | XP_016882647.1 | 3 | 825 | |||
GenBank mRNA | BC105047.1 | - | 2 | 460 | 8144050 | |
BC105049.1 | - | 2 | 460 | 8144052 | ||
D87920.1 | - | 2 | 617 | |||
U66088.1 | - | 2 | 745 |