Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003095.2 | NP_003086.1 | 2 | 174 | ||
GenBank mRNA | AK311752.1 | - | 2 | 137 | ||
BC002505.2 | - | 2 | 167 | 3051263 | ||
BC063397.1 | - | 2 | 124 | 5176590 | ||
BC128452.1 | - | 2 | 77 | 40123237 | ||
BC128453.1 | - | 2 | 76 | 40123239 | ||
BU861031.1 | - | 2 | 177 | |||
CR456751.1 | - | 1 | 31 | |||
X85372.1 | - | 2 | 146 |