Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017371.4 | NP_001017371.3 | 5 | 2227 | ||
NM_001172712.1 | NP_001166183.1 | 7 | 2569 | |||
NM_003111.4 | NP_003102.1 | 7 | 2578 | |||
GenBank mRNA | AB209334.1 | - | 4 | 1691 | ||
AK304199.1 | - | 7 | 2150 | |||
AY070137.1 | - | 5 | 2227 | |||
AY441957.1 | - | 7 | 2578 | |||
AY441958.1 | - | 5 | 1917 | |||
BC042945.2 | - | 4 | 781 | |||
BC126414.1 | - | 7 | 2100 | 8992130 | ||
BC143928.1 | - | 7 | 2091 | 9052446 | ||
CR749323.1 | - | 2 | 3784 | |||
M97191.2 | - | 5 | 1838 | |||
X68560.1 | - | 5 | 1796 |