Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142522.1 | NP_001135994.1 | 1 | 167 | ||
GenBank mRNA | AJ420499.1 | - | 1 | 27 | ||
AK055221.1 | - | 1 | 141 | |||
BI838832.1 | - | 1 | 198 |