siRNA Details
Gene Symbol: UBTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076683.1 | NP_001070151.1 | 7 | 843 | ||
NM_001076684.2 | NP_001070152.1 | 7 | 929 | |||
NM_014233.3 | NP_055048.1 | 7 | 929 | |||
NR_045058.1 | 7 | 1126 | ||||
XM_006722059.3 | XP_006722122.1 | 7 | 968 | |||
XM_006722060.2 | XP_006722123.1 | 6 | 712 | |||
XM_006722061.2 | XP_006722124.1 | 7 | 796 | |||
XM_017024995.1 | XP_016880484.1 | 7 | 977 | |||
XM_017024996.1 | XP_016880485.1 | 7 | 929 | |||
XM_017024997.1 | XP_016880486.1 | 7 | 1121 | |||
XM_017024998.1 | XP_016880487.1 | 7 | 837 | |||
XM_017024999.1 | XP_016880488.1 | 6 | 712 | |||
XM_017025000.1 | XP_016880489.1 | 7 | 881 | |||
XM_017025001.1 | XP_016880490.1 | 7 | 929 | |||
XM_017025002.1 | XP_016880491.1 | 7 | 809 | |||
XM_017025003.1 | XP_016880492.1 | 7 | 819 | |||
XM_017025004.1 | XP_016880493.1 | 7 | 811 | |||
GenBank mRNA | AK291733.1 | - | 7 | 754 | ||
AK292518.1 | - | 7 | 1126 | |||
AK292577.1 | - | 7 | 1014 | |||
AK296917.1 | - | 2 | 153 | |||
AK298008.1 | - | 5 | 455 | |||
AK308321.1 | - | 6 | 906 | |||
BC031423.1 | - | 6 | 867 | |||
BC042297.1 | - | 7 | 834 | 4509695 | ||
M61724.1 | - | 1 | 24 | |||
X53390.1 | - | 6 | 780 | |||
X53461.1 | - | 6 | 780 | |||
X56687.1 | - | 7 | 812 |
