Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024712.3 | NP_078988.2 | 19 | 2118 | ||
GenBank mRNA | AI955864.1 | - | 2 | 428 | ||
AK023886.1 | - | 18 | 1882 | |||
AK056923.1 | - | 18 | 2081 | |||
AK300976.1 | - | 19 | 2132 | |||
BC015524.1 | - | 9 | 928 | |||
BC034410.1 | - | 16 | 1730 |