Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG13 SIRNA Assays ›
Gene Symbol:
ALG13
Gene Name:
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases:
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location:
Chr.X: 111680741 - 111764164 on Build GRCh38
UniGene ID:
Hs.443061
Species:
Human
siRNA ID
HSS188235
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039210.4
NP_001034299.3
3
335
NM_001099922.2
NP_001093392.1
3
394
NM_001168385.2
NP_001161857.1
3
394
NM_001257230.1
NP_001244159.1
3
290
NM_001257231.1
NP_001244160.1
3
408
NM_001257234.1
NP_001244163.1
3
363
NM_001257235.2
NP_001244164.1
3
363
NM_001257237.1
NP_001244166.1
3
467
NM_001257239.2
NP_001244168.1
3
382
NM_001257240.2
NP_001244169.1
3
467
NM_001257241.2
NP_001244170.1
3
408
NM_001324290.1
NP_001311219.1
3
400
NM_001324291.1
NP_001311220.1
3
290
NM_001324292.1
NP_001311221.1
3
394
NM_001324293.1
NP_001311222.1
3
467
NM_001324294.1
NP_001311223.1
3
278
NM_018466.5
NP_060936.1
3
394
NR_033125.2
3
290
XM_006724693.2
XP_006724756.1
3
767
XM_006724697.3
XP_006724760.2
3
667
XM_006724698.3
XP_006724761.2
3
792
XM_011531028.2
XP_011529330.1
3
768
XM_011531030.1
XP_011529332.1
3
766
XM_011531031.1
XP_011529333.1
3
467
XM_011531032.1
XP_011529334.1
3
273
XM_011531033.2
XP_011529335.1
3
313
XM_017029846.1
XP_016885335.1
3
769
XM_017029847.1
XP_016885336.1
3
768
XM_017029848.1
XP_016885337.1
3
766
XM_017029849.1
XP_016885338.1
3
767
XM_017029850.1
XP_016885339.1
3
766
XM_017029851.1
XP_016885340.1
3
769
GenBank mRNA
AF220051.1
-
3
325
AK300394.1
-
3
232
AK302729.1
-
3
335
AK302890.1
-
3
311
AK304712.1
-
3
284
AK311608.1
-
3
274
AK312229.1
-
3
333
AK316306.1
-
3
336
AK316522.1
-
3
411
AL524204.3
-
3
213
BC005336.1
-
3
327
3996922
BE747995.1
-
3
323
BF239206.1
-
3
362
BF240663.1
-
3
180
BF699152.1
-
3
396
BG621042.1
-
2
177
BM759764.1
-
3
376
BQ007420.1
-
2
340
CB268871.1
-
3
341
DA830058.1
-
3
458
DB292609.1
-
3
467
DC400909.1
-
3
278
DN992019.1
-
3
462
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
transferase
Gene Ontology Categories:
Function(s)
Process(es)
dolichol-linked oligosaccharide biosynthetic process
proteolysis
N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0