Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153366.3 | NP_699197.3 | 7 | 1851 | ||
GenBank mRNA | AJ619977.1 | - | 7 | 1677 | ||
AK122605.1 | - | 7 | 1847 | |||
AY243503.1 | - | 7 | 1677 | |||
BC030816.1 | - | 7 | 1851 |