Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOVL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104558.1 | NP_001098028.1 | 7 | 691 | ||
NM_001297617.1 | NP_001284546.1 | 10 | 1089 | |||
NM_001297618.1 | NP_001284547.1 | 8 | 696 | |||
NM_024930.2 | NP_079206.2 | 8 | 742 | |||
XM_005248606.4 | XP_005248663.1 | 7 | 1306 | |||
XM_005248607.3 | XP_005248664.1 | 8 | 1019 | |||
XM_006714695.3 | XP_006714758.1 | 9 | 798 | |||
XM_011543651.2 | XP_011541953.1 | 9 | 988 | |||
XM_017009885.1 | XP_016865374.1 | 8 | 679 | |||
XM_017009886.1 | XP_016865375.1 | 10 | 918 | |||
XM_017009887.1 | XP_016865376.1 | 10 | 1109 | |||
GenBank mRNA | AB181393.1 | - | 8 | 683 | ||
AK027216.1 | - | 6 | 512 | |||
AL137506.1 | - | 3 | 173 | |||
BC094792.1 | - | 10 | 1030 | |||
BC130310.1 | - | 7 | 639 | 40146341 | ||
BC130312.1 | - | 7 | 639 | 40146343 |