Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VN1R10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045612.1 | 1 | 933 | |||
GenBank mRNA | AF336874.1 | - | 1 | 894 | ||
AF352327.1 | - | 1 | 912 | |||
BC107050.2 | - | 1 | 895 | 40004452 |