Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM234A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284497.1 | NP_001271426.1 | 9 | 1464 | ||
NM_032039.3 | NP_114428.1 | 9 | 1471 | |||
NR_104317.1 | 9 | 1471 | ||||
XM_005255622.1 | XP_005255679.1 | 9 | 1245 | |||
XM_005255623.2 | XP_005255680.1 | 9 | 1166 | |||
XM_006720957.1 | XP_006721020.1 | 9 | 1188 | |||
XM_011522691.1 | XP_011520993.1 | 10 | 1238 | |||
XM_011522692.1 | XP_011520994.1 | 9 | 1364 | |||
XM_017023760.1 | XP_016879249.1 | 9 | 1581 | |||
XM_017023761.1 | XP_016879250.1 | 10 | 1354 | |||
XM_017023762.1 | XP_016879251.1 | 10 | 1340 | |||
XM_017023763.1 | XP_016879252.1 | 9 | 1345 | |||
XM_017023764.1 | XP_016879253.1 | 9 | 1218 | |||
XM_017023765.1 | XP_016879254.1 | 9 | 1139 | |||
GenBank mRNA | AK057165.1 | - | 9 | 1215 | ||
AK294581.1 | - | 9 | 1183 | |||
AL136542.1 | - | 9 | 1184 | |||
BC010521.1 | - | 7 | 942 | |||
BC013047.1 | - | 8 | 1164 | 3857041 | ||
BC032112.2 | - | 8 | 1164 | 4844625 |