Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134435.2 | NP_001127907.1 | 5 | 681 | ||
NM_001323027.1 | NP_001309956.1 | 5 | 1070 | |||
NM_001323028.1 | NP_001309957.1 | 5 | 893 | |||
NM_001323029.1 | NP_001309958.1 | 6 | 981 | |||
NM_001323030.1 | NP_001309959.1 | 6 | 758 | |||
NM_032263.4 | NP_115639.1 | 6 | 960 | |||
XM_017007316.1 | XP_016862805.1 | 6 | 950 | |||
XM_017007317.1 | XP_016862806.1 | 5 | 681 | |||
GenBank mRNA | AK021729.1 | - | 2 | 505 | ||
AK027224.1 | - | 5 | 679 | |||
AL136889.1 | - | 6 | 944 | |||
BC016156.1 | - | 6 | 908 | |||
CR533515.1 | - | 3 | 535 |