Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASMTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173473.1 | NP_001166944.1 | 9 | 1170 | ||
NM_001173474.1 | NP_001166945.1 | 8 | 1207 | |||
NM_004192.3 | NP_004183.2 | 9 | 1255 | |||
XM_005274434.3 | XP_005274491.1 | 10 | 1361 | |||
XM_011546185.2 | XP_011544487.1 | 7 | 1003 | |||
XM_017029919.1 | XP_016885408.1 | 8 | 1051 | |||
GenBank mRNA | AK001090.1 | - | 9 | 1151 | ||
AK090498.1 | - | 8 | 1159 | |||
AK225770.1 | - | 9 | 1177 | |||
AK297805.1 | - | 8 | 1088 | |||
AK299231.1 | - | 9 | 1173 | |||
AK301844.1 | - | 9 | 1164 | |||
BC002508.2 | - | 9 | 1141 | 3051194 | ||
BC010089.2 | - | 9 | 1139 | 2988425 |