Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199328.2 | NP_955360.1 | 1 | 324 | ||
GenBank mRNA | AK022269.1 | - | 1 | 247 | ||
AK223512.1 | - | 1 | 218 | |||
AY358707.1 | - | 1 | 194 | |||
BC020866.1 | - | 1 | 207 | 4594155 | ||
BC058004.1 | - | 1 | 173 | 4606880 | ||
DA629490.1 | - | 1 | 324 | |||
GQ472207.1 | - | 1 | 207 |