Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303508.1 | NP_001290437.1 | 5 | 1384 | ||
GenBank mRNA | AB219123.1 | - | 4 | 791 | ||
AK025181.1 | - | 4 | 1673 | |||
BC112212.1 | - | 4 | 782 | 8327680 | ||
BC113633.1 | - | 4 | 770 | 8322685 | ||
CR749372.1 | - | 5 | 1385 |