Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAL3ST1 SIRNA Assays ›
Gene Symbol:
GAL3ST1
Gene Name:
galactose-3-O-sulfotransferase 1
Gene Aliases:
CST
Chromosome Location:
Chr.22: 30554635 - 30574588 on Build GRCh38
UniGene ID:
Hs.17958
Species:
Human
siRNA ID
HSS190387
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GAL3ST1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318103.1
NP_001305032.1
3
362
NM_001318104.1
NP_001305033.1
4
472
NM_001318105.1
NP_001305034.1
3
423
NM_001318106.1
NP_001305035.1
4
548
NM_001318107.1
NP_001305036.1
4
536
NM_001318108.1
NP_001305037.1
4
533
NM_001318109.1
NP_001305038.1
3
307
NM_001318110.1
NP_001305039.1
4
440
NM_001318111.1
NP_001305040.1
3
330
NM_001318112.1
NP_001305041.1
3
276
NM_001318113.1
NP_001305042.1
4
386
NM_001318114.1
NP_001305043.1
3
279
NM_001318115.1
NP_001305044.1
4
405
NM_001318116.1
NP_001305045.1
5
528
NM_004861.2
NP_004852.1
4
459
XM_011530518.2
XP_011528820.1
3
1897
XM_011530522.2
XP_011528824.1
3
2013
XM_011530524.1
XP_011528826.1
3
366
XM_011530528.1
XP_011528830.1
3
267
XM_017029096.1
XP_016884585.1
3
483
XM_017029097.1
XP_016884586.1
3
1781
XM_017029098.1
XP_016884587.1
4
1906
GenBank mRNA
AK313097.1
-
3
384
BC014649.1
-
4
366
4564703
BC019077.2
-
3
261
4562507
BG419785.1
-
4
389
BM923857.1
-
4
472
BP220526.1
-
3
302
BP276494.1
-
3
276
CR456350.1
-
3
304
D88667.1
-
4
434
DA271385.1
-
4
418
DA370553.1
-
5
509
DA371478.1
-
3
317
DA372454.1
-
3
353
DA376354.1
-
4
498
DA379889.1
-
4
427
DA381062.1
-
4
438
DA381099.1
-
3
276
DA382951.1
-
4
535
DA384904.1
-
4
548
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
protein N-linked glycosylation
galactosylceramide biosynthetic process
spermatogenesis
myelination
galactosylceramide sulfotransferase activity
sulfotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0