Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178898.4 | NP_849229.2 | 5 | 1324 | ||
GenBank mRNA | AK040541.1 | - | 5 | 1390 | ||
AK045736.1 | - | 5 | 1313 | |||
AK133549.1 | - | 5 | 1325 | |||
AK162106.1 | - | 5 | 1251 | |||
BC090657.1 | - | 5 | 1266 | |||
BC096536.1 | - | 4 | 1034 | |||
BC119280.1 | - | 5 | 1202 | 8734029 | ||
BC120581.1 | - | 5 | 1203 | 40129504 | ||
BC144752.1 | - | 5 | 1203 | 9053300 |