Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199446.1 | NP_955517.1 | 5, 6 | 537 | ||
GenBank mRNA | AK089493.1 | - | 5, 6 | 486 | ||
AK165815.1 | - | 5, 6 | 532 | |||
BC053105.1 | - | 5, 6 | 537 | 6409425 |