Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133966.2 | NP_598727.1 | 5 | 1563 | ||
GenBank mRNA | AK003477.1 | - | 2 | 688 | ||
AK033477.1 | - | 5 | 1591 | |||
AK089463.1 | - | 5 | 1643 | |||
AK154832.1 | - | 5 | 1643 | |||
BC013550.1 | - | 5 | 1563 | 4222283 |