Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018743.4 | NP_061213.2 | 11 | 2109 | ||
GenBank mRNA | AF015811.1 | - | 8 | 785 | ||
AF406611.1 | - | 10 | 1620 | |||
AK045235.1 | - | 11 | 2003 | |||
AK077561.1 | - | 11 | 1993 | |||
AK083589.1 | - | 11 | 2003 | |||
AK161270.1 | - | 11 | 1972 | |||
AK162033.1 | - | 8 | 914 | |||
AK167876.1 | - | 11 | 2018 | |||
AK169257.1 | - | 11 | 2003 | |||
AY489184.1 | - | 10 | 1178 | |||
BC031767.1 | - | 10 | 1463 | 4457846 | ||
BC057860.1 | - | 8 | 783 |