Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145926.3 | NP_666038.3 | 7 | 1084 | ||
GenBank mRNA | AB053218.1 | - | 7 | 786 | ||
AK155336.1 | - | 7 | 1085 | |||
AK177701.1 | - | 2 | 98 | |||
AK186986.1 | - | 2 | 98 | |||
AK190141.1 | - | 98 | ||||
AK191887.1 | - | 2 | 98 | |||
AK195601.1 | - | 2 | 98 | |||
AK199018.1 | - | 2 | 98 | |||
AK199371.1 | - | 97 | ||||
AK202109.1 | - | 2 | 98 | |||
AK204039.1 | - | 2 | 98 | |||
AK205336.1 | - | 2 | 98 | |||
AK210581.1 | - | 2 | 98 | |||
AK213265.1 | - | 2 | 98 | |||
AK217144.1 | - | 2 | 98 | |||
BC026638.1 | - | 2 | 192 | 4984907 | ||
BC031613.1 | - | 1 | 62 | 4216720 | ||
CX205743.1 | - | 4 | 344 |