Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145926.3 | NP_666038.3 | 12 | 1679 | ||
GenBank mRNA | AB053218.1 | - | 12 | 1381 | ||
AK155336.1 | - | 12 | 1680 | |||
BC026638.1 | - | 7 | 787 | 4984907 | ||
BC031613.1 | - | 6 | 657 | 4216720 | ||
CX206051.1 | - | 4 | 438 |