Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177325.3 | NP_796299.2 | 5 | 986 | ||
GenBank mRNA | AK030545.1 | - | 4 | 892 | ||
AK129350.1 | - | 5 | 940 | |||
AK134365.1 | - | 5 | 970 | |||
AK154063.1 | - | 5 | 973 | |||
BC031531.1 | - | 5 | 959 | 4014819 |