Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170694.1 | NP_001164165.1 | 8 | 883 | ||
NM_134083.4 | NP_598844.3 | 8 | 878 | |||
GenBank mRNA | AK031242.1 | - | 8 | 844 | ||
AK031248.1 | - | 8 | 849 | |||
AK147784.1 | - | 9 | 884 | |||
AK160881.1 | - | 9 | 1070 | |||
BC003224.1 | - | 6 | 676 | 3482729 | ||
JN086338.1 | - | 7 | 799 | |||
JN086339.1 | - | 8 | 1037 |