Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148933.1 | NP_683735.1 | 7 | 1632 | ||
GenBank mRNA | AK033598.1 | - | 7 | 1452 | ||
AK172688.1 | - | 8 | 1555 | |||
BC030719.1 | - | 7 | 1632 | 4501966 | ||
BC030720.1 | - | 7 | 1581 | 4505508 |