Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163529.1 | NP_001157001.1 | 6 | 804 | ||
NM_033615.2 | NP_291093.2 | 6 | 804 | |||
GenBank mRNA | AB059632.1 | - | 6 | 804 | ||
AB059633.1 | - | 6 | 804 | |||
AF386072.1 | - | 2 | 168 | |||
AF472524.1 | - | 6 | 536 | |||
AK048367.1 | - | 2 | 108 | |||
AK053825.1 | - | 6 | 780 | |||
AY054984.1 | - | 6 | 504 | |||
AY382193.1 | - | 6 | 535 | |||
AY382194.1 | - | 6 | 535 | |||
AY953137.1 | - | 1 | 79 | |||
BC138139.1 | - | 6 | 527 | 8861158 |