Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029546.2 | NP_083822.1 | 16 | 2044 | ||
GenBank mRNA | AK088254.1 | - | 16 | 2045 | ||
AK088983.1 | - | 16 | 2045 | |||
BC031787.1 | - | 5 | 531 | |||
BC112325.1 | - | 16 | 2010 | 5355810 |