Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053208.4 | NP_444438.2 | 2 | 1108 | ||
GenBank mRNA | AF340231.1 | - | 1 | 850 | ||
AF453879.1 | - | 2 | 1030 | |||
AJ310547.1 | - | 2 | 1079 | |||
AK075582.1 | - | 2 | 1060 | |||
AK152056.1 | - | 2 | 1069 | |||
AK152223.1 | - | 2 | 1066 | |||
AK159206.1 | - | 2 | 1061 | |||
AK166661.1 | - | 3 | 974 | |||
AK167477.1 | - | 2 | 1061 | |||
BC023299.1 | - | 2 | 1042 | 5135579 | ||
BC051436.1 | - | 1 | 826 | 6312638 | ||
BC086764.1 | - | 2 | 1047 | 5685311 |